Learn More
AdipoGen anti-CD38 (human), mAb (AT1),Monoclonal Antibody.
SDP
Supplier:  AdipoGen ANC187020
Encompass_Preferred
anti-CD38 (human), mAb (AT1), AdipoGen Life Sciences. Monoclonal Antibody. Isotype: Mouse IgG1. Clone: AT1. Applications: FACS. 50 mM Sodium Phosphate pH 7.5, 100 mM Potassium Chloride, 150mM NaCl, 0.5% Gentamicin sulfate Human CD38 is a type II transmembrane glycoprotein with bifunctional ectoenzyme activity catalyzing both synthesis and hydrolysis of cyclic ADP ribose. CD38 is involved in lymphocyte activation and adhesion to endothelium and has been identified as a counter-receptor of CD31.
Catalog No. 50-595-210