Learn More
AdipoGen anti-CD40 (mouse), mAb (FGK45) (preservative free)
SDP
Supplier:  AdipoGen AG20B0036PFC500
Encompass_Preferred
anti-CD40 (mouse), mAb (FGK45) (preservative free), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes mouse CD40. Isotype: Rat IgG2a. Clone: FGK45. Applications: FACS, FUNC. Liquid. In PBS. CD40 belongs to the TNF-receptor superfamily and is essential in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation. The interaction of CD40-CD40L is necessary for amyloid-beta-induced microglial activation, and thus is thought to be an early event in Alzheimer disease pathogenesis. CD40 is constitutively expressed by antigen presenting cells, including dendritic cells, B cells and macrophages. Consistent with its widespread expression on normal cells, CD40 is also expressed on a wide range of tumor cells, including non-Hodgkin's and Hodgkin's lymphomas, myeloma and some carcinomas including nasopharynx, bladder, cervix, kidney and ovary.
Catalog No. 50-463-425