Learn More
Eurofins Discoverx Anti-CD47 antibody, 100 µL. (92-1341)
Supplier:  Eurofins Discoverx 921341
Encompass
CD47 is an antiphagocytic ligand expressed by most cell types that binds the inhibitory receptor signal regulatory protein alpha (SIRPα) on phagocytic cells, sending a 'don't eat me signal' to the phagocyte. Provided as 40μg of antibody in 100 μL buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9850