Learn More
ECM Bioscience CD63 (Extracellular region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CM0401
Encompass_Preferred
Clone (M040) was generated from a proprietary antigen related to the native human CD63 expressed in MeWo melanoma cell line. Accession #P08962. Clone M040 mouse monoclonal antibody has high reactivity to CD63 at the plasma membrane in immunocytochemical assays, and weakly detects a 30-60 kDa* protein on SDS-PAGE "Native" immunoblots of human A431, MeWo, and A549 cells. The antibody does not detect denatured CD63 in western blot assays. Applications: ICC, WB; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = CD63 antigen, Granulophysin, Lysosomal-associated membrane protein 3, LAMP-3, Melanoma-associated antigen ME491, OMA81H, Ocular melanoma-associated antigen, Tetraspanin-30, Tspan-30, MLA1, TSPAN30
Catalog No. 50-206-8644