Learn More
System Biosciences LLC ANTI-CD63 W/ GOAT ANTI-RABBIT AntibodyAnti-CD63 Antibody (rabbit anti-human, mouse) with goat anti-rabbit HRP secondary antibody = 25ul
SDP
Catalog No. NC0571132
Encompass_Preferred
Detect exosomes using SBI’s rigorously validated polyclonal rabbit anti-CD63 antibodies that recognize the human and mouse versions of the protein. When you’re ready to confirm that you’ve isolated exosomes, stick with the exosome experts at SBI. Our polyclonal rabbit anti-CD63 antibodies recognize the human and mouse forms of the protein, and are a great choice for general exosome detection, especially when used in conjunction with antibodies for other exosome markers such as the tetraspanins CD9 and CD81, as well as the heat shock protein HSP70.
Catalog No. NC0571132 Supplier System Biosciences LLC Supplier No. EXOABCD63A1
Detect exosomes using SBI’s rigorously validated polyclonal rabbit anti-CD63 antibodies that recognize the human and mouse versions of the protein. When you’re ready to confirm that you’ve isolated exosomes, stick with the exosome experts at SBI. Our polyclonal rabbit anti-CD63 antibodies recognize the human and mouse forms of the protein, and are a great choice for general exosome detection, especially when used in conjunction with antibodies for other exosome markers such as the tetraspanins CD9 and CD81, as well as the heat shock protein HSP70.