Learn More
ECM Bioscience CD73/NT5E Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CM0641
Encompass_Preferred
Clone (M064) was generated from a proprietary antigen related to the mature form of CD73 expressed in the NCI-H1299 lung cancer cell line. Accession #P21589. Clone M064 mouse monoclonal antibody detects a 65 kDa* protein on SDS-PAGE "Native" immunoblots of human H1299, H1915, and MDA-MB-231 carcinomas. This antibody weakly detects denatured CD73. The antibody works for native western blot, immunoprecipitation, and ELISA. Applications: WB, E, IP; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = 5'-nucleotidase, NT5E, NTE, CD73, NT5
Catalog No. 50-206-8648