Learn More
Sino Biological Anti-CD79B Antibody, Mouse Monoclonal 100 µL
Catalog No. 5000704958
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD79B (rh CD79B; Catalog#29750-H08H; NP_000617.1; Met1-Asp159). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-04958 Supplier Sino Biological Supplier No. 29750MM03100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD79B (rh CD79B; Catalog#29750-H08H; NP_000617.1; Met1-Asp159). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.