Learn More
Biolegend Brilliant Violet 785™ anti-human CD86
Supplier:  Biolegend 305442
Encompass_Preferred
Brilliant Violet 785™ anti-human CD86 [IT2.2]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Common Marmoset, Cotton-topped Tamarin, Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1166965