Learn More
FUJIFILM Biosciences Anti CD9 Rat Monoclonal Antibo
Catalog No. 5000450158
Encompass_Preferred
CD9 is a cell surface glycoprotein which belongs to the tetraspanin superfamily TM4SF and is found on the surface of exosomes This is known to form a complex with integrins and other TM4SF can modulate cell adhesion and migration Also this induces platelet activation and aggregation CD9 is also found on the surface of exosomes This product is a rat monoclonal antibody against CD9 and conjugated with biotin High specificity for human CD9Biotin-conjugated for enhanced detection flexibility
Catalog No. 50-004-50158 Supplier FUJIFILM Biosciences Supplier No. 01728211
CD9 is a cell surface glycoprotein which belongs to the tetraspanin superfamily TM4SF and is found on the surface of exosomes This is known to form a complex with integrins and other TM4SF can modulate cell adhesion and migration Also this induces platelet activation and aggregation CD9 is also found on the surface of exosomes This product is a rat monoclonal antibody against CD9 and conjugated with biotin High specificity for human CD9Biotin-conjugated for enhanced detection flexibility