Learn More
ABclonal Technology CDC42 Polyclonal Antibody, 100UL, Validated in:WB,IHC
SDP
Supplier:  ABclonal Technology A1188
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 118-191 of human CDC42 (NP_001782.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IHC,IF•Swiss Prot: P60953•Gene ID:998•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-6554