Learn More
ECM Bioscience Cdk1 (Tyr-15)[conserved site], phospho-specific Mouse Monoclonal Antibody, ECM Biosciences
Catalog No. 502068655
Encompass_Preferred
Clone M231 was generated from a phospho-Cdk1 (Tyr-15) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Tyr-15 in human Cdk1. This sequence is conserved in rat and mouse Cdk1, and is highly conserved in other Cdks, including Cdk2, Cdk3, Cdk5, and Cdk6. Accession #P06493. The antibody detects a 34 kDa* band corresponding to Cdk1 on SDS-PAGE immunoblots of human SYF and HeLa cells, and this band is removed after alkaline phosphatase treatment. The antibody may also detect Tyr-15 phosphorylation in Cdk2, Cdk3, Cdk5, and Cdk6. Applications: WB, E; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Cdc2
Catalog No. 50-206-8655 Supplier ECM Bioscience Supplier No. CM2311
Add to Cart
Add to Cart
Clone M231 was generated from a phospho-Cdk1 (Tyr-15) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Tyr-15 in human Cdk1. This sequence is conserved in rat and mouse Cdk1, and is highly conserved in other Cdks, including Cdk2, Cdk3, Cdk5, and Cdk6. Accession #P06493. The antibody detects a 34 kDa* band corresponding to Cdk1 on SDS-PAGE immunoblots of human SYF and HeLa cells, and this band is removed after alkaline phosphatase treatment. The antibody may also detect Tyr-15 phosphorylation in Cdk2, Cdk3, Cdk5, and Cdk6. Applications: WB, E; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Cdc2