Learn More
IBL America Anti-Certolizumab pegol (Cimzia) ELISA
Supplier:  IBL America TM09035
Encompass_Preferred
The drug Certolizumab pegol (trade name Cimzia) is a tumor necrosis factor alpha (TNF) blocker and binds to human TNF with a KD of 90pM. Certolizumab pegol is a recombinant, humanized antibody Fab' fragment, with specificity for TNF, conjugated to an approximately 40kDa polyethylene glycol (PEG). The Fab' fragment is manufactured in E. coli and is subsequently subjected to purification and conjugation to PEG2MAL40K, to generate Certolizumab pegol. The ImmunoGuide Antibody to Certolizumab pegol ELISA kit has been designed for the measurement of free antibodies against this drug. It does not detect such antibodies which already are bound to the drug.Enzyme immunoassay for the semi-quantitative determination of free antibodies to Certolizumab pegol (CZP) in serum and plasma. For research use only - not for use in diagnostic procedures.
Catalog No. 50-153-8200