Learn More
Seracare Life Sciences Inc ANTI-CHICKEN IGG (H+L) AB CY5
Supplier:  Seracare Life Sciences Inc 52300379
Encompass_Preferred
Affinity purified polyclonal antibody to Chicken IgG, both heavy and light chains (whole IgG) made in Goat and labeled with Cy5. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-670-72