Learn More
Seracare Life Sciences Inc ANTI-CHICKEN IGG(H+L) AB DL800
Supplier:  Seracare Life Sciences Inc 52300418
Encompass
Affinity purified polyclonal antibody to chicken IgG, both heavy and light chains (whole IgG) made in goat and labeled with DyLight 800. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-671-63