Learn More
Seracare Life Sciences Inc ANTI-CHICKEN IGG (H+L) AB HRP
Supplier:  Seracare Life Sciences Inc 52200373
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Chicken IgG, heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-35