Learn More
Immunology Consultants Monoclonal Mouse anti-Chicken IgY Fc Antibody Clone 7B9, 1.0 mg
SDP
Catalog No. 502414322
Encompass_Preferred
This mouse IgG1 kappa antibody is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with chicken IgY Fc fragment as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4322 Supplier Immunology Consultants Supplier No. MGFC30A7B9
This mouse IgG1 kappa antibody is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with chicken IgY Fc fragment as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator.