Learn More
Immunology Consultants Monoclonal Mouse anti-Chicken IgY Fc Antibody Clone 4C2, 1.0 mg
SDP
Catalog No. 502414321
Encompass_Preferred
Purified Mouse anti-Chicken IgY Fc Antibody. MGFC-30A-4C2 is suitable for detection in sandwich ELISA assay when paired with MGFC-30A-7B9.
Catalog No. 50-241-4321 Supplier Immunology Consultants Supplier No. MGFC30A4C2
Purified Mouse anti-Chicken IgY Fc Antibody. MGFC-30A-4C2 is suitable for detection in sandwich ELISA assay when paired with MGFC-30A-7B9.