Learn More
Immunology Consultants Monoclonal Mouse anti-Chicken IgY Fc Antibody Clone 8A2, 1.0 mg
SDP
Supplier:  Immunology Consultants MGFC30A8A2
Encompass_Preferred
This mouse IgG1 kappa antibody is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with chicken IgY Fc fragment as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4277