Learn More
Immunology Consultants Affinity Purified Goat anti-Hamster (CHO) Lysosomal Phospholipase A2 Antibody, 0.5 mg
SDP
Catalog No. NC2734254
Encompass_Preferred
Goats were immunized with CHO lysosomal phospholipase A2 (LPLA2) of recombinant origin and antiserum were collected. The antibody was immunoaffinity purified off an antigen containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with CHO LPLA2 of recombinant origin and LPLA2 secreted by a null CHO cell line as determined by ELISA and western blot. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. NC2734254 Supplier Immunology Consultants Supplier No. GLPLA65ALY
Goats were immunized with CHO lysosomal phospholipase A2 (LPLA2) of recombinant origin and antiserum were collected. The antibody was immunoaffinity purified off an antigen containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with CHO LPLA2 of recombinant origin and LPLA2 secreted by a null CHO cell line as determined by ELISA and western blot. Optimal working dilutions should be determined experimentally by the investigator.