Learn More
Immunology Consultants Affinity Purified Goat anti-Hamster (CHO) Nucleobindin-2 Antibody, 0.1 mg
SDP
Supplier:  Immunology Consultants GNUC65ALYZ
Encompass_Preferred
Goats were immunized with CHO nucleobindin-2 (NUC) of recombinant origin and antiserum were collected. The antibody was immunoaffinity purified off an antigen containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with CHO NUC of recombinant origin and NUC secreted by a null CHO cell line as determined by ELISA and western blot. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. NC2734223