Learn More
MilliporeSigma Anti-Citrulline (Modified) Detection Kit
Supplier:  MilliporeSigma 17347B
Encompass
The Anti-Citrulline (Modified) Detection Kit provides reagents for the modification and detection of citrulline-containing proteins by Western blot (immublot) analysis.
Catalog No. NC1335055