Learn More
Abcam ANTI CLAUDIN 10 ANTIBODY Key features and detailsRabbit Polyclonal to Claudin 10Suitable for ELISA, WB, IHC-PReacts with HumanIsotype IgG
Supplier:  Abcam AB52234100UG
Encompass_Preferred
ANTI CLAUDIN 10 ANTIBODY Key features and detailsRabbit Polyclonal to Claudin 10Suitable for ELISA, WB, IHC-PReacts with HumanIsotype IgG
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2354720