Learn More
FUJIFILM Biosciences Anti Claudin-5 Rat Monoclonal
Catalog No. 5000450149
Encompass_Preferred
Claudin-5 is a tetra-transmembrane protein with approximately 23kDa belongs to a member of Claudin family It functions as a key component in blood-brain barrier This product is a rat monoclonal antibody which specifically recognizes Claudin-5 and has neutralizing activity have neutralizing activity which reduces barrier function in blood-brain barrier model Specific to Claudin-5 no cross-reactivity with Claudin-1 467 Validated in BBB model for neutralizing function
Catalog No. 50-004-50149 Supplier FUJIFILM Biosciences Supplier No. 01428101
Claudin-5 is a tetra-transmembrane protein with approximately 23kDa belongs to a member of Claudin family It functions as a key component in blood-brain barrier This product is a rat monoclonal antibody which specifically recognizes Claudin-5 and has neutralizing activity have neutralizing activity which reduces barrier function in blood-brain barrier model Specific to Claudin-5 no cross-reactivity with Claudin-1 467 Validated in BBB model for neutralizing function