Learn More
Abcam Anti-Claudin 18.2 antibody [EPR19202-244] Blocking peptide, 1MG
Supplier:  Abcam AB3002411MG
Encompass_Preferred
Anti-Claudin 18.2 antibody [EPR19202-244] Blocking peptide is a Synthetic peptide. >90% purity.
Protein/ Peptide/ Enzyme Synonyms: UNQ778/PRO1572, CLDN18, Claudin-18
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2097342