Learn More
Immunology Consultants anti-Clostridium difficile Toxin A Antibody (Mouse - Clone #3B12) - Monoclonal
SDP
Catalog No. 5000435379
Encompass_Preferred
Clone 3B12 provides high‑specificity detection of C. difficile Toxin A, supporting ELISA, Western blotting, and antigen‑capture assays. Its strong binding affinity and low nonspecific signal make it valuable for toxin characterization, diagnostic assay development, and studies of host–pathogen interactions. MACD‑55A‑3B12 offers consistent performance across complex matrices, enabling reliable quantification of ToxA in research and quality‑control workflows
Catalog No. 50-004-35379 Supplier Immunology Consultants Supplier No. MACD55A3B12
Clone 3B12 provides high‑specificity detection of C. difficile Toxin A, supporting ELISA, Western blotting, and antigen‑capture assays. Its strong binding affinity and low nonspecific signal make it valuable for toxin characterization, diagnostic assay development, and studies of host–pathogen interactions. MACD‑55A‑3B12 offers consistent performance across complex matrices, enabling reliable quantification of ToxA in research and quality‑control workflows