Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Anti-Clumping Agent

Catalog No. 0010057AE
Change view
Click to view available options
Quantity:
20 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
00-100-57AE 20 mL
00-100-57DG 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 00-100-57AE Supplier Gibco™ Supplier No. 0010057AE
Add to Cart
Edge
Add to Cart

GIBCO™ Anti-Clumping Agent is a concentrated liquid reagent that reduces cell clumping to assist in attaining higher viable cell densities in suspension culture.

GIBCO™ Anti-Clumping Agent is a concentrated liquid reagent that reduces cell clumping to assist in attaining higher viable cell densities in suspension culture. Anti-Clumping Agent is a gentle, animal origin-free, chemically defined formulation that contains no proteins, enzymes, hydrolysates, or components of unknown composition.

  • Reduced cell clumping in suspension culture
  • Higher viable cell densities
  • Animal origin-free and chemically defined components
  • Convenient packaging with a larger size now available
  • Customization available to configure packaging and delivery

Simple and Gentle Cell Dissociation

Cell lines adapted to suspension culture can revert to a more adherent or aggregated 'clumping' state. When this occurs, culture condition can be greatly improved by adding a gentle reagent to reduce clumping and adhesion instead of the harsh enzymes often found in cell dissociation products. Anti-Clumping Agent is a simple, gentle, concentrated solution which, when added to culture media (usually titrated between a 1:100 and 1:1000 dilution), can reduce cell clumping significantly.

Reduced Clumping, Increased Viability, Improved Protein Expression

Reduced cell clumping achieved through use of Anti-Clumping Agent is illustrated below (Fig 1). See also the data demonstrating improved viable cell density (Fig 2). Higher viable cell density contributes to the stabilization of culture conditions for better protein yield.

For actual cell dissociation, we recommend our animal origin-free TrypLE™ products.

Use Anti-Clumping Agent after Transfection

Anti-Clumping Agent is not recommended in all suspension cell culture conditions. For example, when creating a cell line using lipid cationic transfection, Anti-Clumping Agent can interfere with the transfection process, so in this case we recommend that Anti-Clumping Agent be used after transfection.

Animal Origin-free and Chemically Defined Components

Because Anti-Clumping Agent is made with only animal origin free and chemically defined components, you can be assured that the risk of adding adventitious agents to your culture system is minimized. Avoiding additives in your system that are not well defined can lead to more robust results from batch to batch.

Convenient and Customizable Packaging

Previously, Anti-Clumping Agent was only available in a 20 mL bottle. However, it is now available in a more economical and convenient 100 mL bottle. For even larger packaging, you can custom configure this product by clicking on the 'Customize this Product' link under the 'Add to Cart' button above or by emailing us at CustomMedia@lifetech.com.

Additional Information and Support

Please review the Manuals and Protocols provided below for additional information on successful use of this product. Direct any unanswered questions to our Technical Services group.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

Content And Storage Store at 2 to 8°C
Cell Line CHO, Insect Cells, 293 (HEK)
Cell Type Mammalian Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type Suspension Cell Culture
Form Liquid
Product Type Anti-clumping Reagent
Format Bottle(s)
Product Line Gibco
Quantity 20 mL
Shipping Condition Room Temperature
Source Animal origin-free
Show More Show Less
How much Anti-clumping agent should I use?

Use anti-clumping agent at a dilution of 1:250 to 1:1000 in culture medium. Test for impact on the cell culture and reduce concentration as the cell clumping is reduced.

Are there any recommendations for preventing or dispersing cell clumps in a suspension culture?

When cells meant to be grown in suspension are grown in static culture, they may form clumps. These clumps will severely limit transfection efficiency and protein expression. It is suggested that FreeStyle 293 cells in FreeStyle media and CHO-S cells in CD-CHO or CHO-SFM are grown in agitated suspension to reduce the appearance of clumps. However, if clumps do form, you can try the following protocol to select for cells that don't form clumps:

- Transfer cells into an appropriate size centrifuge tube that will hold the entire cell suspension.
- Allow cells to sit undisturbed for about 5 minutes. The time can vary depending on the specific cell line. Larger cell clumps will settle to the bottom of the tube.
- Collect cells from the upper portion of the tube to passage into a new flask.

My suspension cells are clumping together. What is going on?

There are a number of factors that can contribute to this:<

1) Presence of calcium and magnesium ions. Wash cells in a balanced salt solution without calcium and magnesium. Gently pipette cells to obtain a single cell suspension.

2) Mycoplasma contamination. Segregate culture and test for mycoplasma infection. Clean hood and incubator. If culture is contaminated, discard.

3) Cell lysis and release of DNA resulting from overdigestion with proteolytic enzymes. Treat cells with DNase I.

4) Certain suspension cell type try to aggregate when they are not under optimal agitation speed and optimization is required.

5) Certain suspension cell type try to aggregate in bioreactors and it is recommended to use Anti-Clumping Agent (Cat. No. 0010057AE).

What reagents do you offer for cell dissociation, and what are the main differences between them?

Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.