Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Anti-Clumping Agent
GREENER_CHOICE

Catalog No. 0010057DG
Change view
Click to view available options
Quantity:
20 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
00-100-57DG 100 mL
00-100-57AE 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 00-100-57DG Supplier Gibco™ Supplier No. 0010057DG
  • PRODUCT PROMOTION
    $427.55
    $365.42 / Each of 1

    Save $62.13

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GIBCO™ Anti-Clumping Agent is a concentrated liquid reagent that reduces cell clumping to assist in attaining higher viable cell densities in suspension culture.

GIBCO™ Anti-Clumping Agent is a concentrated liquid reagent that reduces cell clumping to assist in attaining higher viable cell densities in suspension culture. Anti-Clumping Agent is a gentle, animal origin-free, chemically defined formulation that contains no proteins, enzymes, hydrolysates, or components of unknown composition.

  • Reduced cell clumping in suspension culture
  • Higher viable cell densities
  • Animal origin-free and chemically defined components
  • Convenient packaging with a larger size now available
  • Customization available to configure packaging and delivery

Simple and Gentle Cell Dissociation

Cell lines adapted to suspension culture can revert to a more adherent or aggregated 'clumping' state. When this occurs, culture condition can be greatly improved by adding a gentle reagent to reduce clumping and adhesion instead of the harsh enzymes often found in cell dissociation products. Anti-Clumping Agent is a simple, gentle, concentrated solution which, when added to culture media (usually titrated between a 1:100 and 1:1000 dilution), can reduce cell clumping significantly.

Reduced Clumping, Increased Viability, Improved Protein Expression

Reduced cell clumping achieved through use of Anti-Clumping Agent is illustrated below (Fig 1). See also the data demonstrating improved viable cell density (Fig 2). Higher viable cell density contributes to the stabilization of culture conditions for better protein yield.

For actual cell dissociation, we recommend our animal origin-free TrypLE™ products.

Use Anti-Clumping Agent after Transfection

Anti-Clumping Agent is not recommended in all suspension cell culture conditions. For example, when creating a cell line using lipid cationic transfection, Anti-Clumping Agent can interfere with the transfection process, so in this case we recommend that Anti-Clumping Agent be used after transfection.

Animal Origin-free and Chemically Defined Components

Because Anti-Clumping Agent is made with only animal origin free and chemically defined components, you can be assured that the risk of adding adventitious agents to your culture system is minimized. Avoiding additives in your system that are not well defined can lead to more robust results from batch to batch.

Convenient and Customizable Packaging

Previously, Anti-Clumping Agent was only available in a 20 mL bottle. However, it is now available in a more economical and convenient 100 mL bottle. For even larger packaging, you can custom configure this product by clicking on the 'Customize this Product' link under the 'Add to Cart' button above or by emailing us at CustomMedia@lifetech.com.

Additional Information and Support

Please review the Manuals and Protocols provided below for additional information on successful use of this product. Direct any unanswered questions to our Technical Services group.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

Content And Storage Store at 2 to 8°C
Cell Line CHO, Insect Cells, 293 (HEK)
Cell Type Mammalian Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type Suspension Cell Culture
Form Liquid
Product Type Anti-clumping Reagent
Format Bottle(s)
Product Line Gibco
Quantity 100 mL
Shipping Condition Room Temperature
Source Animal origin-free
Show More Show Less
What reagents do you offer for cell dissociation, and what are the main differences between them?

Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.