Learn More
FUJIFILM Biosciences Anti-Clumping Supplement
Catalog No. NC1206117
Encompass
Part of the BalanCD system of media and supplements, the Anti-Clumping Supplement is an animal-component- and enzyme-free supplement to cell culture medium, designed to reduce cell aggregation for various cell types in suspension culture.
Catalog No. NC1206117 Supplier FUJIFILM Biosciences Supplier No. 9115050ML
Part of the BalanCD system of media and supplements, the Anti-Clumping Supplement is an animal-component- and enzyme-free supplement to cell culture medium, designed to reduce cell aggregation for various cell types in suspension culture.