Learn More
Charles River Labs Cell Solutions Anti-CMV Specific CD8+ T Cells, HLA-B*35 restricted, 1 to 2 Million cells/vial, Cryopreserved
Supplier:  Charles River Labs Cell Solutions ASTC1048
Encompass
This T cell population specifically recognizes an HLA-B35 restricted CMV pp65 peptide (IPSINVHHY) as shown by tetramer binding as well as IFN-gamma secretion assays.
Catalog No. 50-225-6101
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov