Learn More
ACROBiosystems Monoclonal Anti-c-Myc Tag Antibody, Europium Chelate, 20,000 Tests
Catalog No. 5000807435
Encompass_Preferred
The Monoclonal Anti-c-Myc Tag Antibody Europium chelate is a tool for TR-FRET assays, designed for the detection of c-Myc-tagged proteins. This antibody is covalently conjugated with Eu3+ chelate to function as a donor in proximity-based fluorescence assays.
  • Specifically designed for TR-FRET applications
  • Enables detection through 665 nm signal emission upon 620 nm donor excitation
  • Compatible with fluorescently labeled acceptors
  • Validated for binding c-Myc-tagged proteins
  • Supplied as powder for versatile storage
Catalog No. 50-008-07435 Supplier ACROBiosystems Supplier No. FRTTD0720000TESTS
The Monoclonal Anti-c-Myc Tag Antibody Europium chelate is a tool for TR-FRET assays, designed for the detection of c-Myc-tagged proteins. This antibody is covalently conjugated with Eu3+ chelate to function as a donor in proximity-based fluorescence assays.
  • Specifically designed for TR-FRET applications
  • Enables detection through 665 nm signal emission upon 620 nm donor excitation
  • Compatible with fluorescently labeled acceptors
  • Validated for binding c-Myc-tagged proteins
  • Supplied as powder for versatile storage