Learn More
Bioworld Anti-Coatomer Protein Antibody (Rabbit Polyclonal IgG), 200 μg
Supplier:  Bioworld 203121141
Encompass_Preferred
Anti-Coatomer Protein Antibody (Rabbit Polyclonal IgG)Cat#20312114DetailsHost:RabbitIsotype:IgGClass:PolyclonalImmunogen:Synthetic peptide corresponding toHuman and Rat Coatomer ProteinConjugate:UnconjugatedForm InformationForm:LiquidConcentration:2 mg/mLPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:4°C, long term aliquot and store at -20°C. Avoid freeze thaw cycles.Usage RecommendationsWestern Blot (WB):1:3000-1:5000ELISA:1:150-1:400Immunoprecipitation (IP):1:350 - 1:500
Catalog No. 50-198-037