Learn More
Selleck Chemical LLC Anti-Complement C2-1MG-25
SDP
Catalog No. 503138351
Encompass_Preferred
Anti-Complement C2 is a monoclonal antibody that inhibits complement factor C2 It inhibits complement in a disease model for multifocal motor neuropathy MW 146 12 kD
Catalog No. 50-313-8351 Supplier Selleck Chemical LLC Supplier No. A27011MG 25PC
Anti-Complement C2 is a monoclonal antibody that inhibits complement factor C2 It inhibits complement in a disease model for multifocal motor neuropathy MW 146 12 kD