Learn More
ECM Bioscience Coronin-1B (Ser-2), phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CP2621
Encompass_Preferred
This antibody was affinity purified using phosho-Coronin-1B (Ser-2) peptide (without carrier). The antibody detects a 60 kDa* band corresponding to Coronin-1B on SDS-PAGE immunoblots of human A431 or HeLa cells treated with Calyculin A. This band is not observed in control cells and the band observed in Calyculin A-treated cells can be removed with lambda phosphatase treatment. The immunogen was Coronin-1B (Ser-2) synthetic peptide (coupled to carrier protein) corresponds to amino acids surrounding serine 2 in human coronin-1B. This sequence has high homology to the conserved site in rat and mouse coronin-1B and in coronin-1A. However, this sequence has low homology to the conserved region in other coronins. Accession #Q9BR76. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Coronin2, Coro1B
Catalog No. 50-206-8681