Learn More
ABclonal Technology CRADD Polyclonal Antibody, 100UL, Validated in:WB
SDP
Supplier:  ABclonal Technology A1124
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of human RAIDD/RAIDD/CRADD (NP_003796.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse•Validated Applications:WB•Swiss Prot: P78560•Gene ID:8738•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-6504