Learn More
ECM Bioscience Crk II (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CM3321
Encompass_Preferred
Clone (M332) was generated from a recombinant sequence containing the C-terminal half of mouse Crk II. This sequence has high homology to human and rat Crk II. Accession #Q64010. This antibody detects a 40 kDa* protein corresponding to the molecular mass of Crk II on SDS-PAGE immunoblots of rat PC12 and human Jurkat cells. Applications: WB, E, IP; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal, protein A purified antibody is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at -20°C. Stable for 1 year. Other Names = cCrk, c-CRK
Catalog No. 50-206-8660