Learn More
Sino Biological Anti-CSNK1E/CK1 epsilon Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000749314
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CSNK1E fragment, and purified by antigen affinity chromatography.
Catalog No. 50-007-49314 Supplier Sino Biological Supplier No. 200159T34100
Produced in rabbits immunized with E. coli-derived Human CSNK1E fragment, and purified by antigen affinity chromatography.