Learn More
Eurofins Discoverx Anti-CTLA4 Antibody [14D3], 100 µg. (92-1319)
Supplier:  Eurofins Discoverx 921319
Encompass
CTLA4, or CTLA-4 is also known as cytotoxic T-lymphocyte-associated protein 4 or CD152. This monoclonal antibody binds human CTLA4, which functions as a immune checkpoint receptor that downregulates the immune system. CTLA4 is expressed on activated T cells and binds the B7 family members, CD80 (B7-1) and CD86 (B7-2), and the protein restricts the progression of T cells to an activated state by inhibiting IL-2 secretion and cellular proliferation. Provided as 100 μg of antibody in buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9832