Learn More
Medchemexpress LLC Anti-CYR61 CCN1 Anti 5mg | 5mg
SDP
Catalog No. 5000324282
Encompass_Preferred
Anti-CYR61/CCN1 Antibody is a CHO-expressed human antibody that targets CYR61/CCN1 The Anti-CYR61/CCN1 Antibody contains huIgG1 heavy chain and hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for Anti-CYR61/CCN1 Antibody can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24282 Supplier Medchemexpress LLC Supplier No. HYP9903965MG
Anti-CYR61/CCN1 Antibody is a CHO-expressed human antibody that targets CYR61/CCN1 The Anti-CYR61/CCN1 Antibody contains huIgG1 heavy chain and hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for Anti-CYR61/CCN1 Antibody can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)