Learn More
Aviva Systems Biology Corp D930005D10RIK Antibody - C-terminal region (ARP37588_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37588_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against D930005D10RIK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse D930005D10RIK. Alternate names: AI536456, D930005D10RikApplicationWB. Species reactivity: Mouse
Catalog No. 50-014-53822