Learn More
ABclonal Technology DAPK1 Polyclonal Antibody, 100UL, Validated in:WB, IHC
SDP
Supplier:  ABclonal Technology A5741
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 1230-1430 of human DAP Kinase 1 (DAPK1) (NP_004929.2).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IHC•Swiss Prot: P53355•Gene ID:1612•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-8068