Learn More
Seracare Life Sciences Inc ANTI-DEER IGG (H+L) AB FITC
Supplier:  Seracare Life Sciences Inc 52300329
Encompass_Preferred
Affinity purified polyclonal antibody to Deer IgG, both heavy and light chains (whole IgG) made in Rabbit and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
Catalog No. 50-669-15