Learn More
Seracare Life Sciences Inc ANTI-DEER IGG (H+L) ANTIBODY
Supplier:  Seracare Life Sciences Inc 52100209
Encompass
Affinity purified polyclonal antibody to Deer IgG, both heavy and light chains (whole IgG) made in Rabbit. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-668-50