Learn More
Sino Biological Anti-Der f 2 Antibody 100 µL
Catalog No. 5000786540
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dermatophagoides farinae DERF2 ( Catalog#69029-A07E). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-86540 Supplier Sino Biological Supplier No. 69029MM73100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dermatophagoides farinae DERF2 ( Catalog#69029-A07E). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.