Learn More
STEMCELL Technologies Anti-Dextran Antibody, Clone DX1, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60026
Encompass
Clone DX1 is a mouse monoclonal antibody that reacts with dextran, a natural polysaccharide consisting of linear chains of alpha-1,6-linked D-glucopyranose residues with short side chains (mostly 1 - 2 glucose units) 1,3- or 1,4-linked to the backbone of the biopolymer. Verified for use with cell separation, ELISA, and western blotting assays, use this antibody in applications requiring the detection of dextran. This antibody can be used to detect cells labelled with dextran-coated nanoparticles during StemSep™ and EasySep™ cell separation procedures or for other applications that require the detection of dextran. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human T Cell Enrichment Kit (Catalog #19051) and EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5125