Learn More
Med Vet International Contains 90 gr. Kaolin and 4 gr. Pectin
Catalog No. 502188404
Encompass_Preferred
A time-tested oral formula for use as an aid in the treatment of noninfectious diarrhea in horses, cattle, dogs and cats. Combines pectin with kaolin (also known as china clay) in a palatable flavored vehicle. Each fluid ounce (2 tablespoonfuls) contains 90 gr. Kaolin and 4 gr. Pectin.Products are not always manufacturer specific. Actual product appearance may differ from website image.
Catalog No. 50-218-8404 Supplier Med Vet International Supplier No. PHKAOLIN
A time-tested oral formula for use as an aid in the treatment of noninfectious diarrhea in horses, cattle, dogs and cats. Combines pectin with kaolin (also known as china clay) in a palatable flavored vehicle. Each fluid ounce (2 tablespoonfuls) contains 90 gr. Kaolin and 4 gr. Pectin.Products are not always manufacturer specific. Actual product appearance may differ from website image.