Learn More
Metrohm USA Antidiffusion valve for antidiffusion buret tips
SDP
Catalog No. NC0265267 Shop All Metrohm USA Products
Encompass
For 6.1541.XXX and 6.1543.130, 6.1543.140, 6.1543.150, 6.1543.160, 6.1543.200 antidiffusion buret tips. Outer diameter 4.9mm ; Length 6mm
Catalog No. NC0265267 Supplier Metrohm USA Supplier No. 62726090
For 6.1541.XXX and 6.1543.130, 6.1543.140, 6.1543.150, 6.1543.160, 6.1543.200 antidiffusion buret tips. Outer diameter 4.9mm ; Length 6mm