Learn More
Aviva Systems Biology Corp DKFZP564J0863 Antibody - middle region (ARP47163_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47163_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DKFZP564J0863. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DKFZP564J0863. Alternate names: ATL3, DKFZP564J0863ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53830