Learn More
Aviva Systems Biology Corp DKFZp686E2433 Antibody - N-terminal region (ARP51740_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51740_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DKFZp686E2433. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DKFZp686E2433. Alternate names: DKFZp686E2433ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53668