Learn More
Aviva Systems Biology Corp DKFZP761C169 Antibody - N-terminal region (ARP32876_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32876_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DKFZP761C169. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human DKFZP761C169. Alternate names: GPBP, SSH6, VASCULIN, DKFZP761C169ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53813