Learn More
Aviva Systems Biology Corp DKFZp779O175 Antibody - N-terminal region (ARP50424_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50424_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DKFZp779O175. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DKFZp779O175. Alternate names: DKFZp686K21248, FLJ27459, MGC129866, MGC129867, DKFZp779O175ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-53631